accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 328000-16-4

OMgBr

CAS RN 328000-16-4

IUPAC Name: magnesium(2+) 2-(benzyloxy)benzen-1-ide bromide
Synonyms: magnesium(2+) 2-(benzyloxy)benzen-1-ide bromide
Molecular Weight (g/mol): 287.44
Molecular Formula: C13H11BrMgO
InChi Key: WHVIUIJRSBHYEZ-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].C(OC1=CC=CC=[C-]1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results