CAS RN 32795-84-9
CAS RN 32795-84-9
IUPAC Name:
7-bromotetraphene
Synonyms:
7-bromotetraphene
Molecular Weight (g/mol):
307.19
Molecular Formula:
C18H11Br
InChi Key:
LGRNWCDRODWMOH-UHFFFAOYSA-N
SMILES:
BrC1=C2C=CC3=CC=CC=C3C2=CC2=CC=CC=C12
1
–
3
of
3
results
1
7-Bromobenz[a]anthracene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-Bromobenz[a]anthracene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
7-Bromobenz[a]anthracene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.