accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3279-76-3

CH3OHN

CAS RN 3279-76-3

IUPAC Name: 6-methyl-1,2-dihydropyridin-2-one
Synonyms: 6-methyl-1H-pyridin-2-one
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: JEAVIRYCMBDJIU-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=O)N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)