accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3ClCl

CAS RN 32768-54-0

IUPAC Name: 1,2-dichloro-3-methylbenzene
Synonyms: 1,2-dichloro-3-methylbenzene
Molecular Weight (g/mol): 161.03
Molecular Formula: C7H6Cl2
InChi Key: GWLKCPXYBLCEKC-UHFFFAOYSA-N
SMILES: CC1=CC=CC(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)