accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 327183-32-4

OO

CAS RN 327183-32-4

IUPAC Name: 3,4-dihydro-2H-1-benzopyran-8-carbaldehyde
Synonyms: 3,4-dihydro-2H-1-benzopyran-8-carbaldehyde
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: ZYKTXRDSAFQAAM-UHFFFAOYSA-N
SMILES: O=CC1=C2OCCCC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results