accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 32679-02-0

IUPAC Name: bis(acetonitrile) cycloocta-1,5-diene tetrafluoroboranuide rhodium
Synonyms: 1,5-cyclooctadiene bis(acetonitrile) rhodium tetrafluoroborate
Molecular Weight (g/mol): 380
Molecular Formula: C12H18BF4N2Rh
InChi Key: ZIBLHOBPBGAKNV-UHFFFAOYSA-N
SMILES: [Rh].CC#N.CC#N.F[B-](F)(F)F.C1CC=CCCC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results