accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 32677-01-3

CH3H3CCH3OOOOCH3H3CCH3H2NHCl

CAS RN 32677-01-3

IUPAC Name: 1,5-di-tert-butyl 2-aminopentanedioate hydrochloride
Synonyms: 1,5-di-tert-butyl 2-aminopentanedioate hydrochloride
Molecular Weight (g/mol): 295.8
Molecular Formula: C13H26ClNO4
InChi Key: LFEYMWCCUAOUKZ-UHFFFAOYNA-N
SMILES: Cl.CC(C)(C)OC(=O)CCC(N)C(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)