accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 32673-41-9

HONHNHCl

CAS RN 32673-41-9

IUPAC Name: hydrogen (1H-imidazol-5-yl)methanol chloride
Synonyms: hydrogen 3H-imidazol-4-ylmethanol chloride
Molecular Weight (g/mol): 134.56
Molecular Formula: C4H7ClN2O
InChi Key: WFNASTYGEKUMIY-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OCC1=CN=CN1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)