accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3259-03-8

OBrOCH3

CAS RN 3259-03-8

IUPAC Name: 1-(2-bromoethoxy)-2-ethoxybenzene
Synonyms: 1-(2-bromoethoxy)-2-ethoxybenzene
Molecular Weight (g/mol): 245.12
Molecular Formula: C10H13BrO2
InChi Key: IOYHGBZPUZBUTJ-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1OCCBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results