accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CCH3SiH3CCH3ONO

CAS RN 325486-37-1

IUPAC Name: 1-benzyl-3-{[(tert-butyldimethylsilyl)oxy]methyl}piperidin-4-one
Synonyms: 1-benzyl-3-{[(tert-butyldimethylsilyl)oxy]methyl}piperidin-4-one
Molecular Weight (g/mol): 333.55
Molecular Formula: C19H31NO2Si
InChi Key: WOYQEVXRQBRUAL-UHFFFAOYNA-N
SMILES: CC(C)(C)[Si](C)(C)OCC1CN(CC2=CC=CC=C2)CCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results