accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 325465-93-8

CH3OOOPOHOOOHNa(E/Z)

CAS RN 325465-93-8

IUPAC Name: sodium 3-(hydrogen phosphonatooxy)-2-hydroxypropyl octadec-9-enoate
Synonyms: sodium 3-(hydrogen phosphonatooxy)-2-hydroxypropyl octadec-9-enoate
Molecular Weight (g/mol): 458.51
Molecular Formula: C21H40NaO7P
InChi Key: XGRLSUFHELJJAB-UHFFFAOYNA-M
SMILES: [Na+].CCCCCCCCC=CCCCCCCCC(=O)OCC(O)COP(O)([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results