accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 325461-35-6

H3CCH3CH3OOOCH3CH3CH3O

CAS RN 325461-35-6

IUPAC Name: 2,5-bis[(3,7-dimethyloctyl)oxy]benzene-1,4-dicarbaldehyde
Synonyms: 2,5-bis[(3,7-dimethyloctyl)oxy]benzene-1,4-dicarbaldehyde
Molecular Weight (g/mol): 446.67
Molecular Formula: C28H46O4
InChi Key: JMACLBZCZVVLDQ-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)CCOC1=CC(C=O)=C(OCCC(C)CCCC(C)C)C=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results