accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 32503-34-7

H3CNCH3H3CCH3OHSOOO

CAS RN 32503-34-7

IUPAC Name: tetrahexylazanium sulfate
Synonyms: tetrahexylazanium sulfate
Molecular Weight (g/mol): 450.74
Molecular Formula: C24H52NO4S
InChi Key: RULHPTADXJPDSN-UHFFFAOYSA-L
SMILES: [O-]S([O-])(=O)=O.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results