CAS RN 32385-06-1
CAS RN 32385-06-1
IUPAC Name:
4-amino-6-methoxy-2-methyloxan-3-ol hydrochloride
Synonyms:
4-amino-6-methoxy-2-methyloxan-3-ol hydrochloride
Molecular Weight (g/mol):
197.66
Molecular Formula:
C7H16ClNO3
InChi Key:
JVYGBUAXDCLPMP-UHFFFAOYNA-N
SMILES:
Cl.COC1CC(N)C(O)C(C)O1
1
–
2
of
2
results
1
Methyl Alpha-L-Daunosamine Hydrochloride (Alpha:Beta approx. 85:15), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methyl alpha-L-Daunosamine Hydrochloride (alpha:beta approx. 85:15), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.