accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3235-82-3

H3COONHO

CAS RN 3235-82-3

IUPAC Name: 4-(2-ethoxy-2-oxoethyl)morpholin-4-ium
Synonyms: 4-(2-ethoxy-2-oxoethyl)morpholin-4-ium
Molecular Weight (g/mol): 174.22
Molecular Formula: C8H16NO3
InChi Key: SITMDWHJQROIPF-UHFFFAOYSA-O
SMILES: CCOC(=O)C[NH+]1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results