accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 32324-19-9

CH3SOONCO

CAS RN 32324-19-9

IUPAC Name: 2-methylbenzene-1-sulfonyl isocyanate
Synonyms: 2-methylbenzenesulfonyl isocyanate
Molecular Weight (g/mol): 197.21
Molecular Formula: C8H7NO3S
InChi Key: HEBTZZBBPUFAFE-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1S(=O)(=O)N=C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)