accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 323-09-1

F

CAS RN 323-09-1

IUPAC Name: 2-fluoronaphthalene
Synonyms: naphthalene, 2-fluoro-
Molecular Weight (g/mol): 146.16
Molecular Formula: C10H7F
InChi Key: BAGQBTMEEISJLK-UHFFFAOYSA-N
SMILES: FC1=CC=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results