accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3229-73-0

CH3H3CNH2OCH3H3CNO

CAS RN 3229-73-0

IUPAC Name: 1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxamide
Synonyms: 1-hydroxy-2,2,5,5-tetramethylpyrrole-3-carboxamide
Molecular Weight (g/mol): 184.24
Molecular Formula: C9H16N2O2
InChi Key: GMXOTWPDUBXCMS-UHFFFAOYSA-N
SMILES: CC1(C)C=C(C(N)=O)C(C)(C)N1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results