accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 32249-35-7

H3COOO

CAS RN 32249-35-7

IUPAC Name: methyl 3-cyclopropyl-3-oxopropanoate
Synonyms: methyl 3-cyclopropyl-3-oxopropanoate
Molecular Weight (g/mol): 142.15
Molecular Formula: C7H10O3
InChi Key: RIJWDPRXCXJDPK-UHFFFAOYSA-N
SMILES: COC(=O)CC(=O)C1CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results