accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

IISm

CAS RN 32248-43-4

IUPAC Name: λ²-samarium(2+) diiodide
Synonyms: samarium(II) iodide
Molecular Weight (g/mol): 404.17
Molecular Formula: I2Sm
InChi Key: UAWABSHMGXMCRK-UHFFFAOYSA-L
SMILES: [I-].[I-].[Sm++]
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)