accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3222-49-9

CH3NOHO

CAS RN 3222-49-9

IUPAC Name: 5-methylpyridine-3-carboxylic acid
Synonyms: 5-methylpyridine-3-carboxylic acid
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: DJDHHXDFKSLEQY-UHFFFAOYSA-N
SMILES: CC1=CN=CC(=C1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)