accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 32190-42-4

NN

CAS RN 32190-42-4

IUPAC Name: 2,2'-bipyridine
Synonyms: bipyridyl α,α dipyridyl dipyridyl, 2,2' α,α'-dipyridyl α,α'-bipyridine 2,2-bipyridyl bpy dipyridyl, 2,2 2,2 bipyridine α,α-dipyridyl
Molecular Weight (g/mol): 156.19
Molecular Formula: C10H8N2
InChi Key: ROFVEXUMMXZLPA-UHFFFAOYSA-N
SMILES: C1=CC=C(N=C1)C1=CC=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results