accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 321744-26-7

CH3OO(R)(S)OHNOOH3CCH3H3C

CAS RN 321744-26-7

IUPAC Name: 1-tert-butyl 2-methyl (2R,4S)-4-hydroxypiperidine-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-methyl (2R,4S)-4-hydroxypiperidine-1,2-dicarboxylate
Molecular Weight (g/mol): 259.3
Molecular Formula: C12H21NO5
InChi Key: RNMVWSAJMIKMDY-DTWKUNHWSA-N
SMILES: COC(=O)[C@H]1C[C@@H](O)CCN1C(=O)OC(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)