accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CONO

CAS RN 32161-06-1

IUPAC Name: 1-acetylpiperidin-4-one
Synonyms: 1-acetyl-piperidin-4-one
Molecular Weight (g/mol): 141.17
Molecular Formula: C7H11NO2
InChi Key: NNFOVLFUGLWWCL-UHFFFAOYSA-N
SMILES: CC(=O)N1CCC(=O)CC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)