CAS RN 321351-00-2
IUPAC Name:
4-bromo-N1-[3-chloro-4-(2-methylbenzoyl)phenyl]benzene-1,2-diamine
Synonyms:
4-bromo-N1-[3-chloro-4-(2-methylbenzoyl)phenyl]benzene-1,2-diamine
Molecular Weight (g/mol):
415.72
Molecular Formula:
C20H16BrClN2O
InChi Key:
HDCLCHNAEZNGNV-UHFFFAOYSA-N
SMILES:
CC1=CC=CC=C1C(=O)C1=C(Cl)C=C(NC2=C(N)C=C(Br)C=C2)C=C1