CAS RN 32133-82-7
CAS RN 32133-82-7
IUPAC Name:
[2-({diphenyl[(1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-yl)oxy]-λ⁴-sulfanyl}oxy)-1,1,1,3,3,3-hexafluoropropan-2-yl]benzene
Synonyms:
[2-({diphenyl[(1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-yl)oxy]-λ⁴-sulfanyl}oxy)-1,1,1,3,3,3-hexafluoropropan-2-yl]benzene
Molecular Weight (g/mol):
672.53
Molecular Formula:
C30H20F12O2S
InChi Key:
RMIBJVUYNZSLSD-UHFFFAOYSA-N
SMILES:
FC(F)(F)C(OS(OC(C1=CC=CC=C1)(C(F)(F)F)C(F)(F)F)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C(F)(F)F
1
–
3
of
3
results
1
Bis[a,a-bis(trifluoromethyl)benzenemethanolato]diphenylsulfur, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Bis[a,a-bis(trifluoromethyl)benzenemethanolato]diphenylsulfur, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Bis[a,a-bis(trifluoromethyl)benzenemethanolato]diphenylsulfur, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.