accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 321309-24-4

ONHNH2HN

CAS RN 321309-24-4

IUPAC Name: 1H-indole-7-carbohydrazide
Synonyms: 1H-indole-7-carbohydrazide
Molecular Weight (g/mol): 175.19
Molecular Formula: C9H9N3O
InChi Key: NCFMBDUFIJHHAW-UHFFFAOYSA-N
SMILES: NNC(=O)C1=C2NC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results