accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 321-14-2

HOOClHO

CAS RN 321-14-2

IUPAC Name: 5-chloro-2-hydroxybenzoic acid
Synonyms: 5-chlorosalicylic acid 5 csa chlorosalicylic acid
Molecular Weight (g/mol): 172.56
Molecular Formula: C7H5ClO3
InChi Key: NKBASRXWGAGQDP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=CC=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results