accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 32085-88-4

FFO

CAS RN 32085-88-4

IUPAC Name: 3,5-difluorobenzaldehyde
Synonyms: 3,5-difluorobenzaldehyde
Molecular Weight (g/mol): 142.11
Molecular Formula: C7H4F2O
InChi Key: ASOFZHSTJHGQDT-UHFFFAOYSA-N
SMILES: FC1=CC(C=O)=CC(F)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)