accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3205-25-2

H3COHNOO

CAS RN 3205-25-2

IUPAC Name: 1-(2-nitrophenyl)ethan-1-ol
Synonyms: 1-(2-nitrophenyl)ethanol
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: DSDBYQDNNWCLHL-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=CC=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)