accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 320423-50-5

CH3NNNSO

CAS RN 320423-50-5

IUPAC Name: 2-(4-methylpiperazin-1-yl)-1,3-thiazole-5-carbaldehyde
Synonyms: 2-(4-methylpiperazin-1-yl)-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol): 211.28
Molecular Formula: C9H13N3OS
InChi Key: TUJAFVJUJXMFEG-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=NC=C(S1)C=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)