accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OONHNNH2

CAS RN 32016-28-7

IUPAC Name: benzyl 5-amino-1H-pyrazole-4-carboxylate
Synonyms: benzyl 3-amino-2H-pyrazole-4-carboxylate
Molecular Weight (g/mol): 217.23
Molecular Formula: C11H11N3O2
InChi Key: QBEUXDWEKIOSIL-UHFFFAOYSA-N
SMILES: NC1=C(C=NN1)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results