CAS RN 32004-67-4
CAS RN 32004-67-4
IUPAC Name:
2-[(1E)-5-fluoro-2-methyl-1-{[4-(methylsulfanyl)phenyl]methylidene}-1H-inden-3-yl]acetic acid
Synonyms:
[(3E)-6-fluoro-2-methyl-3-{[4-(methylsulfanyl)phenyl]methylidene}inden-1-yl]acetic acid
Molecular Weight (g/mol):
340.41
Molecular Formula:
C20H17FO2S
InChi Key:
LFWHFZJPXXOYNR-RQZCQDPDSA-N
SMILES:
CSC1=CC=C(\C=C2/C(C)=C(CC(O)=O)C3=CC(F)=CC=C23)C=C1