accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 320-88-7

ONOFFFFFF

CAS RN 320-88-7

IUPAC Name: 2-nitro-1,4-bis(trifluoromethyl)benzene
Synonyms: 2-nitro-1,4-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 259.11
Molecular Formula: C8H3F6NO2
InChi Key: BZKVUOHHNCMTLH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)