accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 320-47-8

NOOFFFN

CAS RN 320-47-8

IUPAC Name: 4-nitro-2-(trifluoromethyl)benzonitrile
Synonyms: 4-nitro-2-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 216.12
Molecular Formula: C8H3F3N2O2
InChi Key: AGKQJEFSEQHGTA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=C(C=C1)C#N)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results