CAS RN 320-42-3
CAS RN 320-42-3
IUPAC Name:
4-hydroxy-2-(trifluoromethyl)benzonitrile
Synonyms:
4-hydroxy-2-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol):
187.12
Molecular Formula:
C8H4F3NO
InChi Key:
MDBVIOUGHSKRMT-UHFFFAOYSA-N
SMILES:
OC1=CC(=C(C=C1)C#N)C(F)(F)F
1
–
1
of
1
results
1
4-Hydroxy-2-(trifluoromethyl)benzonitrile, 98%
CAS: 320-42-3 Molecular Formula: C8H4F3NO Molecular Weight (g/mol): 187.121 MDL Number: MFCD06797340 InChI Key: MDBVIOUGHSKRMT-UHFFFAOYSA-N Synonym: 4-hydroxy-2-trifluoromethyl benzonitrile,4-hydroxy-2-trifluoroemthyl benzonitrile,pubchem4807,2-cyano-5-hydroxybenzotrifluoride,4-cyano-3-trifluoromethyl phenol,4-hydroxy-2-trifluoromethyl-benzonitrile,benzonitrile, 4-hydroxy-2-trifluoromethyl,4-hydroxy-2-trifluoromethyl benzenecarbonitrile PubChem CID: 2783153 IUPAC Name: 4-hydroxy-2-(trifluoromethyl)benzonitrile SMILES: C1=CC(=C(C=C1O)C(F)(F)F)C#N