accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31938-07-5

BrN

CAS RN 31938-07-5

IUPAC Name: 2-(3-bromophenyl)acetonitrile
Synonyms: 2-(3-bromophenyl)acetonitrile
Molecular Weight (g/mol): 196.05
Molecular Formula: C8H6BrN
InChi Key: UUZYFBXKWIQKTF-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(CC#N)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results