CAS RN 3190-71-4
CAS RN 3190-71-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(4S)-2,5-Dioxo-4-oxazolidinepropanoic Acid Phenylmethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(4S)-2,5-Dioxo-4-oxazolidinepropanoic Acid Phenylmethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(4S)-2,5-Dioxo-4-oxazolidinepropanoic Acid Phenylmethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.