accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3189-13-7

CH3OHN

CAS RN 3189-13-7

IUPAC Name: 6-methoxy-1H-indole
Synonyms: 6-methoxy-1H-indole
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: QJRWYBIKLXNYLF-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CNC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)