CAS RN 318469-22-6
CAS RN 318469-22-6
IUPAC Name:
[1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
Synonyms:
[1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazol-4-yl]methanol
Molecular Weight (g/mol):
272.23
Molecular Formula:
C12H11F3N2O2
InChi Key:
TVYDMZWMEOZWHY-UHFFFAOYSA-N
SMILES:
CN1N=C(C(CO)=C1OC1=CC=CC=C1)C(F)(F)F