accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31835-06-0

H3COO(S)O(R)OH(S)OH(S)OH(R)OH(S)OHO(R)HO(S)OH(S)OH

CAS RN 31835-06-0

IUPAC Name: (2S,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl decanoate
Synonyms: (2S,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl decanoate
Molecular Weight (g/mol): 496.55
Molecular Formula: C22H40O12
InChi Key: KQXPBHIAPSOIPX-RAGLHBGGSA-N
SMILES: CCCCCCCCCC(=O)O[C@]1(O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@@]1(CO)O[C@H](CO)[C@@H](O)[C@@H]1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)