accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31823-05-9

OOH

CAS RN 31823-05-9

IUPAC Name: (1-benzofuran-5-yl)methanol
Synonyms: 1-benzofuran-5-ylmethanol
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: XSLXZYDPOMAXTM-UHFFFAOYSA-N
SMILES: OCC1=CC=C2OC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results