CAS RN 317854-65-2
CAS RN 317854-65-2
IUPAC Name:
3-formyl-1H-indole-7-carboxylic acid
Synonyms:
3-formyl-1H-indole-7-carboxylic acid
Molecular Weight (g/mol):
189.17
Molecular Formula:
C10H7NO3
InChi Key:
HABYJDPNVBUDGD-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C2NC=C(C=O)C2=CC=C1