accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 3178-22-1

CH3H3CCH3

CAS RN 3178-22-1

IUPAC Name: tert-butylcyclohexane
Synonyms: tert-butylcyclohexane
Molecular Weight (g/mol): 140.27
Molecular Formula: C10H20
InChi Key: XTVMZZBLCLWBPM-UHFFFAOYSA-N
SMILES: CC(C)(C)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results