CAS RN 317318-96-0
CAS RN 317318-96-0
IUPAC Name:
{4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methanol
Synonyms:
{4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methanol
Molecular Weight (g/mol):
273.27
Molecular Formula:
C12H10F3NOS
InChi Key:
SEHCYQHVWYXBGI-UHFFFAOYSA-N
SMILES:
CC1=C(CO)SC(=N1)C1=CC=C(C=C1)C(F)(F)F