accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2NOHO

CAS RN 3167-49-5

IUPAC Name: 6-aminopyridine-3-carboxylic acid
Synonyms: 6-aminopyridine-3-carboxylic acid
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: ZCIFWRHIEBXBOY-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=N1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results