CAS RN 31658-49-8
CAS RN 31658-49-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
17Alpha-Methyl-5Beta-estrane-3Alpha,17Beta-diol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
17alpha-Methyl-5beta-estrane-3alpha,17beta-diol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
17alpha-Methyl-5beta-estrane-3alpha,17beta-diol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.