accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31618-90-3

CH3OPOOSOOCH3OCH3

CAS RN 31618-90-3

IUPAC Name: diethyl [(4-methylbenzenesulfonyl)oxy]methanephosphonate
Synonyms: diethyl [(4-methylbenzenesulfonyl)oxy]methanephosphonate
Molecular Weight (g/mol): 322.31
Molecular Formula: C12H19O6PS
InChi Key: UOEFFQWLRUBDME-UHFFFAOYSA-N
SMILES: CCOP(=O)(COS(=O)(=O)C1=CC=C(C)C=C1)OCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results