accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 316173-57-6

CH3ONNNNNNH2NO

CAS RN 316173-57-6

IUPAC Name: 4-(furan-2-yl)-10-[3-(4-methoxyphenyl)propyl]-3,5,6,8,10,11-hexaazatricyclo[7.3.0.0²,⁶]dodeca-1(9),2,4,7,11-pentaen-7-amine
Synonyms: 4-(furan-2-yl)-10-[3-(4-methoxyphenyl)propyl]-3,5,6,8,10,11-hexaazatricyclo[7.3.0.0²,⁶]dodeca-1(9),2,4,7,11-pentaen-7-amine
Molecular Weight (g/mol): 389.42
Molecular Formula: C20H19N7O2
InChi Key: AEULVFLPCJOBCE-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCCN2N=CC3=C2N=C(N)N2N=C(N=C32)C2=CC=CO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results