accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 31602-08-1

H3COONClClCl

CAS RN 31602-08-1

IUPAC Name: ethyl 4,6,8-trichloroquinoline-3-carboxylate
Synonyms: ethyl 4,6,8-trichloroquinoline-3-carboxylate
Molecular Weight (g/mol): 304.55
Molecular Formula: C12H8Cl3NO2
InChi Key: HAPRHXIRCWGCIG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C2C(Cl)=CC(Cl)=CC2=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)